Learn More
4-Nitrophenol-2,3,5,6-d4, CDN
Product Code. 30320052
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320052
Brand: CDN CDND23560.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Nitrophenol-2,3,5,6-D4
CAS 93951-79-2
Molecular Formula C6 2H4 H N O3
Quantity 0.1 g
Synonym 4-Nitrophenol-2,3,5,6-D4
InChI Formula InChI=1 S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8 H/i1D,2 D,3 D,4 D
SMILES [2 H]c1c([2 H])c(c([2 H])c([2 H])c1O)[N+](=O)[O-]
IUPAC Name 2,3,5,6-tetradeuterio-4-nitrophenol
Molecular Weight (g/mol) 143.13
Formula Weight 143.0521 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity