missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzyl-d6 Chloride, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4-Fluorobenzyl Chloride-D6 |
| CAS | 1219804-13-3 |
| Molecular Formula | C7 D6 Cl F |
| Quantity | 0.1 g |
| Synonym | 4-Fluorobenzyl-d6 Chloride, 4-Fluorobenzyl Chloride-d6, 4-Fluorobenzyl Chloride-D6, 1-(Chloromethyl-d2)-4-fluorobenzene-2,3,5,6-d4, α-Chloro-4-fluorotoluene-d6 |
| InChI Formula | InChI=1 S/C7H6ClF/c8-5-6-1-3-7(9)4-2-6/h1-4 H,5H2/i1D,2 D,3 D,4 D,5D2 |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1F)C([2 H])([2 H])Cl |
| IUPAC Name | 1-[chloro(dideuterio)methyl]-2,3,5,6-tetradeuterio-4-fluorobenzene |
| Molecular Weight (g/mol) | 150.6109 |
| Formula Weight | 150.0519 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction