missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromophenyl Phenyl-d5 Ether, CDN
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
Specifications
Specifications
| Chemical Name or Material | 4-Bromodiphenyl Ether 2',3',4',5',6'-d5 |
| CAS | 93951-83-8 |
| Molecular Formula | C12 2H5 H4 Br O |
| Quantity | 0.1 g |
| Synonym | Benzene-d5, (4-bromophenoxy)- (9 CI), 1-Bromo-4-(pentadeuteriophenoxy)benzene, 4-Bromodiphenyl ether (2',3',4',5',6'-D4), 4-Bromophenyl phenyl ether (phenyl-d5) |
| InChI Formula | InChI=1 S/C12H9BrO/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9 H/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(Oc2ccc(Br)cc2)c([2 H])c1[2 H] |
| IUPAC Name | 1-(4-bromophenoxy)-2,3,4,5,6-pentadeuteriobenzene |
| Molecular Weight (g/mol) | 254.13 |
| Formula Weight | 253.0151 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction