missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminobiphenyl-d9, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | 4-Aminobiphenyl D9 |
| CAS | 344298-96-0 |
| Molecular Formula | C12 2H9 H2 N |
| Quantity | 0.1 g |
| Synonym | [1,1'-Biphenyl-2,2',3,3',4',5,5',6,6'-d9]-4-amine (9 CI), 4-Aminobiphenyl D9, 4-Aminobiphenyl-D9 |
| InChI Formula | InChI=1 S/C12H11N/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9 H,13H2/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2c([2 H])c([2 H])c(N)c([2 H])c2[2 H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)aniline |
| Molecular Weight (g/mol) | 178.28 |
| Formula Weight | 178.1456 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction