missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dihydroxybiphenyl-d8 (rings-d8), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4,4'-Dihydroxybiphenyl-2,2',3,3',5,5',6,6'-D8 |
| CAS | 612480-60-1 |
| Molecular Formula | C12 D8 H2 O2 |
| Quantity | 0.1 g |
| Synonym | [1,1'-Biphenyl-2,2',3,3',5,5',6,6'-d8]-4,4'-diol (9 CI), 4,4'-Dihydroxybiphenyl-d8 (rings-d8), 4,4'-Dihydroxybiphenyl-2,2',3,3',5,5',6,6'-D8, p,p'-Biphenol-d8 |
| InChI Formula | InChI=1 S/C12H10O2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8,13-14 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1O)c2c([2 H])c([2 H])c(O)c([2 H])c2[2 H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)phenol |
| Molecular Weight (g/mol) | 194.2559 |
| Formula Weight | 194.1183 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction