missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dichlorobenzophenone-d8, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4,4'-Dichlorobenzophenone D8 |
| CAS | 1219806-01-5 |
| Molecular Formula | C13 2H8 Cl2 O |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C13H8Cl2O/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10/h1-8 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)c2c([2 H])c([2 H])c(Cl)c([2 H])c2[2 H])c([2 H])c([2 H])c1Cl |
| IUPAC Name | bis(4-chloro-2,3,5,6-tetradeuteriophenyl)methanone |
| Molecular Weight (g/mol) | 259.16 |
| Formula Weight | 258.0454 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction