missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-1-propanol-1,1,2,2,3,3-d6, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-1-propanol-D6 |
| CAS | 284474-43-7 |
| Molecular Formula | C3 D6 H Br O |
| Quantity | 0.1 g |
| Synonym | 1-Propan-1,1,2,2,3,3-d6-ol, 3-bromo- (9 CI), 3-Bromo-1,1,2,2,3,3-hexadeuteriopropan-1-ol, 3-Bromo-1-propanol-1,1,2,2,3,3-d6, 3-Bromo-1-propanol-D6, 1-Bromo-3-hydroxypropane-1,1,2,2,3,3-d6 |
| InChI Formula | InChI=1 S/C3H7BrO/c4-2-1-3-5/h5H,1-3H2/i1D2,2D2,3D2 |
| SMILES | [2 H]C([2 H])(O)C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 3-bromo-1,1,2,2,3,3-hexadeuteriopropan-1-ol |
| Molecular Weight (g/mol) | 145.0281 |
| Formula Weight | 144.0057 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction