missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propylpentanoic-d15 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Valproic-D15 Acid |
| CAS | 362049-65-8 |
| Molecular Formula | C8 D15 H O2 |
| Quantity | 0.1 g |
| Synonym | Pentanoic-2,3,3,4,4,5,5,5-d8 acid, 2-(propyl-d7)- (9 CI), Pentadecadeuterovalproic acid, 2-(Propyl-d7)pentanoic-2,3,3,4,4,5,5,5-d8 acid, Valproic acid-d15, Valproic-D15 acid |
| InChI Formula | InChI=1 S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)/i1D3,2D3,3D2,4D2,5D2,6D2,7 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C(=O)O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,3,4,4,5,5,5-octadeuterio-2-(1,1,2,2,3,3,3-heptadeuteriopropyl)pentanoic acid |
| Molecular Weight (g/mol) | 159.3039 |
| Formula Weight | 159.2092 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction