missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2-Methyl-d3-piperazine-2,3,3,5,5,6,6-d7, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2-Methylpiperazine-D10 |
| CAS | 2363789-19-7 |
| Molecular Formula | C5 D10 H2 N2 |
| Quantity | 0.1 g |
| Synonym | (±)-2-Methyl-d3-piperazine-2,3,3,5,5,6,6-d7, 6-(Methyl-d3)piperazine-2,2,3,3,5,5,6-d7, 2-Methylpiperazine-d10, 2-Methylpiperazine-D10 |
| InChI Formula | InChI=1 S/C5H12N2/c1-5-4-6-2-3-7-5/h5-7 H,2-4H2,1H3/i1D3,2D2,3D2,4D2,5 D |
| SMILES | [2 H]C([2 H])([2 H])C1([2 H])NC([2 H])([2 H])C([2 H])([2 H])NC1([2 H])[2 H] |
| IUPAC Name | 2,2,3,3,5,5,6-heptadeuterio-6-(trideuteriomethyl)piperazine |
| Molecular Weight (g/mol) | 110.2238 |
| Formula Weight | 110.1628 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction