missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptobenzimidazole-4,5,6,7-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-Mercapto-4,5,6,7-d4-benzimidazole |
| CAS | 931581-17-8 |
| Molecular Formula | C7 2H4 H2 N2 S |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C7H6N2S/c10-7-8-5-3-1-2-4-6(5)9-7/h1-4 H,(H2,8,9,10)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2[nH]cnc2c1[2 H] |
| IUPAC Name | 4,5,6,7-tetradeuterio-1 H-benzimidazole-2-thiol |
| Molecular Weight (g/mol) | 154.23 |
| Formula Weight | 154.0503 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction