missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-2-methyl-d3-propionic-3,3,3-d3 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy-2-methyl-d3-propanoic-3,3,3-d3 Acid |
| CAS | 40662-45-1 |
| Molecular Formula | C4 2H6 H2 O3 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C4H8O3/c1-4(2,7)3(5)6/h7H,1-2H3,(H,5,6)/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(O)(C(=O)O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 3,3,3-trideuterio-2-hydroxy-2-(trideuteriomethyl)propanoic acid |
| Molecular Weight (g/mol) | 110.14 |
| Formula Weight | 110.085 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction