missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylphenol-d10, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | 2-Ethylphenol-D10 |
| CAS | 721429-63-6 |
| Molecular Formula | C8 D10 O |
| Quantity | 0.1 g |
| Synonym | Phen-2,3,4,5-d4-ol, 6-(ethyl-d5)- (9 CI), 2-Ethylphenol-D10, 2-Ethylphenol-d10, 2-(Ethyl-d5)phenol-O,3,4,5,6-d5, 1-Ethyl-2-hydroxybenzene-d10, o-Ethylphenol-d10, Phlorol-d10 |
| InChI Formula | InChI=1 S/C8H10O/c1-2-7-5-3-4-6-8(7)9/h3-6,9 H,2H2,1H3/i1D3,2D2,3 D,4 D,5 D,6 D/hD |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c([2 H])c([2 H])c1C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,3,4-tetradeuterio-5-deuteriooxy-6-(1,1,2,2,2-pentadeuterioethyl)benzene |
| Molecular Weight (g/mol) | 132.226 |
| Formula Weight | 132.1359 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction