Learn More
2-Chlorobiphenyl-2',3',4',5',6'-d5, CDN
Product Code. 30321326
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321326
Brand: CDN CDND30110.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chlorobiphenyl-d5
CAS 51624-35-2
Molecular Formula C12 2H5 H4 Cl
Quantity 0.1 g
Synonym 1,1'-Biphenyl-2,3,4,5,6-d5, 2'-chloro- (9 CI), 2-Chlorobiphenyl D5, 2'-Chloro-2,3,4,5,6-pentadeuterio-1,1'-biphenyl
InChI Formula InChI=1 S/C12H9Cl/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9 H/i1D,2 D,3 D,6 D,7 D
SMILES [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2ccccc2Cl
IUPAC Name 1-(2-chlorophenyl)-2,3,4,5,6-pentadeuteriobenzene
Molecular Weight (g/mol) 193.68
Formula Weight 193.0707 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity