missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzaldehyde-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-Chlorobenzaldehyde-3,4,5,6-D4 |
| CAS | 1093351-46-2 |
| Molecular Formula | C7 D4 H Cl O |
| Quantity | 0.1 g |
| Synonym | Benzaldehyde-2,3,4,5-d4, 6-chloro- (ACI), 6-Chlorobenzaldehyde-2,3,4,5-d4 (ACI), 2-Chlorobenzaldehyde-d4, 2-Chlorobenzaldehyde-3,4,5,6-d4, 2-Chlorobenzaldehyde-3,4,5,6-D4 |
| InChI Formula | InChI=1 S/C7H5ClO/c8-7-4-2-1-3-6(7)5-9/h1-5 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C=O)c(Cl)c1[2 H] |
| IUPAC Name | 2-chloro-3,4,5,6-tetradeuteriobenzaldehyde |
| Molecular Weight (g/mol) | 144.5917 |
| Formula Weight | 144.028 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction