missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Naphthalenedicarboxylic-d6 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 2,6-Naphthalenedicarboxylic Acid-D6 |
| CAS | 682321-46-6 |
| Molecular Formula | C12 D6 H2 O4 |
| Quantity | 0.1 g |
| Synonym | 2,6-Naphthalene-1,3,4,5,7,8-d6-dicarboxylic acid (9 CI), 2,6-Naphthalenedicarboxylic-d6 Acid, 2,6-Naphthalenedicarboxylic Acid-D6 |
| InChI Formula | InChI=1 S/C12H8O4/c13-11(14)9-3-1-7-5-10(12(15)16)4-2-8(7)6-9/h1-6 H,(H,13,14)(H,15,16)/i1D,2 D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c2c([2 H])c(C(=O)O)c([2 H])c([2 H])c2c([2 H])c1C(=O)O |
| IUPAC Name | 1,3,4,5,7,8-hexadeuterionaphthalene-2,6-dicarboxylic acid |
| Molecular Weight (g/mol) | 222.2265 |
| Formula Weight | 222.0799 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction