Learn More
2,4,6-Trichlorophenol-3,5-d2, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2,4,6-Trichlorophenol ring-d2 |
| CAS | 93951-80-5 |
| Molecular Formula | C6 2H2 H Cl3 O |
| Quantity | 0.1 g |
| Synonym | Phen-3,5-d2-ol, 2,4,6-trichloro-, 2,4,6-Trichlorophen-3,5-d2-ol, 2,4,6-Trichlorophenol D2, 2,4,6-Trichlorophenol (ring-D2) |
| InChI Formula | InChI=1 S/C6H3Cl3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10 H/i1D,2 D |
| SMILES | [2 H]c1c(Cl)c([2 H])c(Cl)c(O)c1Cl |
| IUPAC Name | 2,4,6-trichloro-3,5-dideuteriophenol |
| Molecular Weight (g/mol) | 199.46 |
| Formula Weight | 197.9375 g/mol |
| Show More |