missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trichloroanisole-d5, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | 2,4,6-Trichloroanisole D5 |
| CAS | 352439-08-8 |
| Molecular Formula | C7 2H5 Cl3 O |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C7H5Cl3O/c1-11-7-5(9)2-4(8)3-6(7)10/h2-3 H,1H3/i1D3,2 D,3 D |
| SMILES | [2 H]c1c(Cl)c([2 H])c(Cl)c(OC([2 H])([2 H])[2 H])c1Cl |
| IUPAC Name | 1,3,5-trichloro-2,4-dideuterio-6-(trideuteriomethoxy)benzene |
| Molecular Weight (g/mol) | 216.5 |
| Formula Weight | 214.972 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction