missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,6-Trichlorophenol-4,5-d2, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2,3,6-Trichlorophenol ring-d2 |
| CAS | 93951-81-6 |
| Molecular Formula | C6 2H2 H Cl3 O |
| Quantity | 0.1 g |
| Synonym | Phen-3,4-d2-ol, 2,5,6-trichloro- (9 CI), 2,3,6-Trichlorophenol-3,4-d2, 3,4-Dideuterio-2,3,6-trichlorophenol, 2,3,6-Trichlorophenol (ring-D2) |
| InChI Formula | InChI=1 S/C6H3Cl3O/c7-3-1-2-4(8)6(10)5(3)9/h1-2,10 H/i1D,2 D |
| SMILES | [2 H]c1c([2 H])c(Cl)c(Cl)c(O)c1Cl |
| IUPAC Name | 2,3,6-trichloro-4,5-dideuteriophenol |
| Molecular Weight (g/mol) | 199.46 |
| Formula Weight | 197.9375 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction