Learn More
2,3,5,6-Tetrachloroaniline-d3, CDN
Product Code. 30316410
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316410
Brand: CDN CDND69110.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,3,5,6-Tetrachloroaniline-D3
CAS 1219806-05-9
Molecular Formula C6 D3 Cl4 N
Quantity 0.1 g
Synonym 2,3,5,6-Tetrachlorobenzene-4-d-amine-d2, 2,3,5,6-Tetrachloroaniline-d3, 2,3,5,6-Tetrachloroaniline-D3
InChI Formula InChI=1 S/C6H3Cl4N/c7-2-1-3(8)5(10)6(11)4(2)9/h1H,11H2/i1D/hD2
SMILES [2 H]N([2 H])c1c(Cl)c(Cl)c([2 H])c(Cl)c1Cl
IUPAC Name 2,3,5,6-tetrachloro-N,N,4-trideuterioaniline
Molecular Weight (g/mol) 233.9252
Formula Weight 231.9208 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity