Learn More
1-Isopropyl-d7-piperazine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Isopropylpiperazine-D7 (isopropyl-D7) |
| CAS | 2089679-76-3 |
| Molecular Formula | C7 D7 H9 N2 |
| Quantity | 0.1 g |
| Synonym | Piperazine, 1-[1-(methyl-d3)ethyl-1,2,2,2-d4]- (ACI), 1-[1-(Methyl-d3)ethyl-1,2,2,2-d4]piperazine (ACI), 1-Isopropyl-d7-piperazine, 1-Isopropylpiperazine-D7 (isopropyl-D7), Isopropylpiperazine-d7, N-(Isopropyl-d7)piperazine |
| InChI Formula | InChI=1 S/C7H16N2/c1-7(2)9-5-3-8-4-6-9/h7-8 H,3-6H2,1-2H3/i1D3,2D3,7 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(N1CCNCC1)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)piperazine |
| Molecular Weight (g/mol) | 135.2585 |
| Formula Weight | 135.1753 g/mol |
| Show More |