missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Dodecane-d25-thiol, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Dodecanethiol-D25 |
| CAS | 869563-00-8 |
| Molecular Formula | C12 D25 H S |
| Quantity | 0.1 g |
| Synonym | 1-Dodecane-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25-thiol (9 CI, ACI), 1-Dodecane-d25-thiol, 1-Dodecanethiol-d25, 1-Dodecanethiol-D25, 1-Dodecyl-d25 mercaptan, Lauryl-d25 mercaptan |
| InChI Formula | InChI=1 S/C12H26S/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])S |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-pentacosadeuteriododecane-1-thiol |
| Molecular Weight (g/mol) | 227.5539 |
| Formula Weight | 227.3324 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction