Learn More
1-Bromododecane-12,12,12-d3, CDN
Click to view available options
Quantity:
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromododecane-12,12,12-D3 |
| CAS | 204259-68-7 |
| Molecular Formula | C12 D3 H22 Br |
| Quantity | 0.1 g |
| Synonym | Dodecane-1,1,1-d3, 12-bromo- (9 CI), 12-Bromododecane-1,1,1-d3, 1-Bromododecane-12,12,12-d3, 1-Bromododecane-12,12,12-D3, Dodecyl bromide-12,12,12-d3, Lauryl bromide-ω-d3 |
| InChI Formula | InChI=1 S/C12H25Br/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-12H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCCCCCCCCCCBr |
| IUPAC Name | 12-bromo-1,1,1-trideuteriododecane |
| Molecular Weight (g/mol) | 252.2494 |
| Formula Weight | 251.1328 g/mol |
| Show More |