Learn More
1-Aminonaphthalene-d9, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | 1-Aminonaphthalene-D9 |
| CAS | 78832-56-1 |
| Molecular Formula | C10 D9 N |
| Quantity | 0.1 g |
| Synonym | 1-Naphthalen-2,3,4,5,6,7,8-d7-amine-d2 (9 CI), 1-Napthylamine-d9, 1-Aminonaphthalene-d9, 1-Aminonaphthalene-D9, 1-Aminonaphthalene-N,N,2,3,4,5,6,7,8-d9 |
| InChI Formula | InChI=1 S/C10H9N/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7 H,11H2/i1D,2 D,3 D,4 D,5 D,6 D,7 D/hD2 |
| SMILES | [2 H]N([2 H])c1c([2 H])c([2 H])c([2 H])c2c([2 H])c([2 H])c([2 H])c([2 H])c12 |
| IUPAC Name | N,N,2,3,4,5,6,7,8-nonadeuterionaphthalen-1-amine |
| Molecular Weight (g/mol) | 152.2406 |
| Formula Weight | 152.13 g/mol |
| Show More |