Learn More
1,3-Propane-2,2-d2-diamine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1,3-Diaminopropane-2,2-D2 |
| CAS | 352438-78-9 |
| Molecular Formula | C3 D2 H8 N2 |
| Quantity | 0.1 g |
| Synonym | 1,3-Propane-2,2-d2-diamine (9 CI), 2,2-Dideuteriopropane-1,3-diamine, 1,3-Diaminopropane-d2, 1,3-Diaminopropane-2,2-D2, Trimethylenediamine-d2 |
| InChI Formula | InChI=1 S/C3H10N2/c4-2-1-3-5/h1-5H2/i1D2 |
| SMILES | [2 H]C([2 H])(CN)CN |
| IUPAC Name | 2,2-dideuteriopropane-1,3-diamine |
| Molecular Weight (g/mol) | 76.1372 |
| Formula Weight | 76.097 g/mol |
| Show More |