Learn More
1,3-Diethylbenzene-d14, CDN
Product Code. 30319761
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319761
Brand: CDN CDND63080.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,3-Diethylbenzene-D14
CAS 1219803-40-3
Molecular Formula C10 D14
Quantity 0.1 g
Synonym 1,3-Di(ethyl-d5)benzene-2,4,5,6-d4, 1,3-Diethylbenzene-d14, 1,3-Diethylbenzene-D14, m-Diethylbenzene-d14
InChI Formula InChI=1 S/C10H14/c1-3-9-6-5-7-10(4-2)8-9/h5-8 H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2,5 D,6 D,7 D,8 D
SMILES [2 H]c1c([2 H])c(c([2 H])c(c1[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC Name 1,2,3,5-tetradeuterio-4,6-bis(1,1,2,2,2-pentadeuterioethyl)benzene
Molecular Weight (g/mol) 148.3044
Formula Weight 148.1974 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity