missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,16-Hexadecanedioic-d28 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Hexadecanedioic Acid-D28 |
| CAS | 130348-90-2 |
| Molecular Formula | C16 D28 H2 O4 |
| Quantity | 0.1 g |
| Synonym | Hexadecanedioic-d28 acid (9 CI), Hexadecanedioic acid-d28, 1,16-Hexadecanedioic-d28 acid, 1,14-Tetradecanedicarboxylic-d28 acid, Hexadecanedioic Acid-D28 |
| InChI Formula | InChI=1 S/C16H30O4/c17-15(18)13-11-9-7-5-3-1-2-4-6-8-10-12-14-16(19)20/h1-14H2,(H,17,18)(H,19,20)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2 |
| SMILES | [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15-octacosadeuteriohexadecanedioic acid |
| Molecular Weight (g/mol) | 314.5795 |
| Formula Weight | 314.3902 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction