Learn More
1,1,1,2-Tetrachloroethane-d2, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1,1,1,2-Tetrachloroethane-D2 |
| CAS | 117164-18-8 |
| Molecular Formula | C2 D2 Cl4 |
| Quantity | 0.1 g |
| Synonym | Ethane-1,1-d2, 1,2,2,2-tetrachloro- (ACI), 1,2,2,2-Tetrachloroethane-1,1-d2 (ACI), Ethane-1,1-d2, tetrachloro- (6 CI), 1,1,1,2-Tetrachloroethane-d2, 1,1,1,2-Tetrachloroethane-D2, 1,1,1,2-Tetrachloroethane-2,2-d2 |
| InChI Formula | InChI=1 S/C2H2Cl4/c3-1-2(4,5)6/h1H2/i1D2 |
| SMILES | [2 H]C([2 H])(Cl)C(Cl)(Cl)Cl |
| IUPAC Name | 1,1,1,2-tetrachloro-2,2-dideuterioethane |
| Molecular Weight (g/mol) | 169.8616 |
| Formula Weight | 167.9036 g/mol |
| Show More |