Learn More
Trifluralin-d14 (di-n-propyl-d14), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Trifluralin D14 (di-n-propyl D14) |
| CAS | 347841-79-6 |
| Molecular Formula | C13 2H14 H2 F3 N3 O4 |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C13H16F3N3O4/c1-3-5-17(6-4-2)12-10(18(20)21)7-9(13(14,15)16)8-11(12)19(22)23/h7-8 H,3-6H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N(c1c(cc(cc1[N+](=O)[O-])C(F)(F)F)[N+](=O)[O-])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N-bis(1,1,2,2,3,3,3-heptadeuteriopropyl)-2,6-dinitro-4-(trifluoromethyl)aniline |
| Molecular Weight (g/mol) | 349.37 |
| Formula Weight | 349.1972 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |