Learn More
Sodium 2-(Propyl-1,1-d2)pentanoate-3,3-d2, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Sodium Valproate-3,3,3',3'-D4 |
| CAS | 2749637-13-4 |
| Molecular Formula | C8 D4 H11 O2 . Na |
| Quantity | 0.05 g |
| Synonym | Sodium 2-(propyl-1,1-d2)pentanoate-3,3-d2, 3,3,3',3'-Tetradeuterovalproic acid sodium salt, 2-(Propyl-1,1-d2)pentanoic-3,3-d2 acid sodium salt, Valproic acid-d4 sodium salt, Valproic-3,3,3',3'-D4 acid sodium salt, Sodium valproate-3,3,3',3'-D4, Sodium valproate-3,3,3',3'-D4 |
| InChI Formula | InChI=1 S/C8H16O2.Na/c1-3-5-7(6-4-2)8(9)10;/h7H,3-6H2,1-2H3,(H,9,10);/q;+1/p-1/i5D2,6D2; |
| SMILES | [Na+].[2 H]C([2 H])(CC)C(C(=O)[O-])C([2 H])([2 H])CC |
| IUPAC Name | sodium;3,3-dideuterio-2-(1,1-dideuteriopropyl)pentanoate |
| Molecular Weight (g/mol) | 170.218 |
| Formula Weight | 170.122 g/mol |
| Show More |