missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Quizalofop-ethyl-d3 (propionate-3,3,3-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Quizalofop-ethylD3(3,3,3-D3) |
| CAS | 1398065-84-3 |
| Molecular Formula | C19 2H3 H14 Cl N2 O4 |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C19H17ClN2O4/c1-3-24-19(23)12(2)25-14-5-7-15(8-6-14)26-18-11-21-17-10-13(20)4-9-16(17)22-18/h4-12 H,3H2,1-2H3/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1)C(=O)OCC |
| IUPAC Name | ethyl 2-[4-(6-chloroquinoxalin-2-yl)oxyphenoxy]-3,3,3-trideuteriopropanoate |
| Molecular Weight (g/mol) | 375.82 |
| Formula Weight | 375.1065 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction