missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphorylcholine-d9 Chloride, Calcium Salt (N,N,N-trimethyl-d9), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Phosphocholine-d9 Chloride Calcium Salt |
| CAS | 344299-43-0 |
| Molecular Formula | C5H4D9CaClNO4P |
| Quantity | 0.05 g |
| Synonym | N,N,N-Trimethyl-2-(phosphonooxy)ethanaminium-d9 Chloride Calcium Salt, Biocalcose-d9, Calcium Phosphorylcholine-d9, Calcium Phosphorylcholine-d9 Chloride, Colifos-d9, Colincalcium-d9, Epafosforil-d9, Fosfocolina-d9, Ipercolin-d9, Isocolin-d9, Mebophos-d9, Merival-d9, Pancholin-d9, Phoscholin-d9; |
| InChI Formula | InChI=1 S/C5H14NO4P.Ca.ClH/c1-6(2,3)4-5-10-11(7,8)9;;/h4-5H2,1-3H3,(H-,7,8,9);;1 H/q;+2;/p-2/i1D3,2D3,3D3;; |
| SMILES | O=P([O-])([O-])OCC[N+](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H].[Ca+2 ].[Cl-] |
| IUPAC Name | calcium 2-(tris(methyl-d3)ammonio)ethyl phosphate chloride |
| Molecular Weight (g/mol) | 266.72 |
| Formula Weight | 266.05 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction