missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecanoic-17,17,18,18,18-d5 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Octadecanoic-17,17,18,18,18-D5 Acid |
| CAS | 211443-83-3 |
| Molecular Formula | C18H31D5O2 |
| Quantity | 0.05 g |
| Synonym | Octadecanoic-17,17,18,18,18-d5 Acid, Octadecanoic-17,17,18,18,18-d5 acid (9 CI), Stearic-17,17,18,18,18-d5 acid, 17,17,18,18,18-Pentadeuterostearic acid, [17,17,18,18,18-d5]-Stearic acid, Stearic acid-17,17,18,18,18-d5, Octadecanoic-17,17,18,18,18-D5 acid |
| InChI Formula | InChI=1 S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i1D3,2D2 |
| SMILES | OC(CCCCCCCCCCCCCCCC([2 H])([2 H])C([2 H])([2 H])[2 H])=O |
| IUPAC Name | octadecanoic-17,17,18,18,18-d5 acid |
| Molecular Weight (g/mol) | 289.51 |
| Formula Weight | 289.3 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido