missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Phenyl-d5-acetyl)glycine, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-(Phenyl-d5-acetyl)glycine |
| CAS | 1189920-31-7 |
| Molecular Formula | C10 D5 H6 N O3 |
| Quantity | 0.05 g |
| Synonym | 2-[[2-(2,3,4,5,6-Pentadeuteriophenyl)acetyl]amino]acetic acid, 2-(2-Phenylacetamido)acetic acid D5 (phenyl-D5), Phenaceturic acid-d5, N-(2-Phenyl-d5-acetyl)glycine, (2-Phenylacetyl)glycine-d5, 2-(2-Phenylacetamido)acetic acid-d5, 2-[[2-(Phenyl-d5)acetyl]amino]acetic acid, N-(Phenyl-d5-acetyl)glycine, N-Phenacetylglycine-d5, Phenacetylglycine-d5, Phenylacetylglycine-d5 |
| InChI Formula | InChI=1 S/C10H11NO3/c12-9(11-7-10(13)14)6-8-4-2-1-3-5-8/h1-5 H,6-7H2,(H,11,12)(H,13,14)/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CC(=O)NCC(=O)O)c([2 H])c1[2 H] |
| IUPAC Name | 2-[[2-(2,3,4,5,6-pentadeuteriophenyl)acetyl]amino]acetic acid |
| Molecular Weight (g/mol) | 198.23 |
| Formula Weight | 198.105 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction