missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Oleoyl-2-amino-1,3-propane-d5-diol, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-Oleoyl-2-amino-1,3-propanediol-1,1,2,3,3-D5 |
| Molecular Formula | C21 D5 H36 N O3 |
| Quantity | 0.05 g |
| Synonym | (9 Z)-N-[2-Hydroxy-1-(hydroxymethyl-d2)ethyl-1,2,2-d3]-9-octadecenamide, N-Oleoyl serinol-d5, N-Oleoyl-2-amino-1,3-propane-d5-diol, N-Oleoyl-2-amino-1,3-propanediol-1,1,2,3,3-D5 |
| InChI Formula | InChI=1 S/C21H41NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(25)22-20(18-23)19-24/h9-10,20,23-24 H,2-8,11-19H2,1H3,(H,22,25)/b10-9-/i18D2,19D2,20 D |
| SMILES | [2 H]C([2 H])(O)C([2 H])(NC(=O)CCCCCCC\C=C/CCCCCCCC)C([2 H])([2 H])O |
| IUPAC Name | (Z)-N-(1,1,2,3,3-pentadeuterio-1,3-dihydroxypropan-2-yl)octadec-9-enamide |
| Molecular Weight (g/mol) | 360.586 |
| Formula Weight | 360.34 g/mol |
| Percent Purity | 98 atom % D, min 96% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction