Learn More
N-Nitrosodimethyl-d6-amine, CDN
Product Code. 30326978
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30326978
Brand: CDN CDND29370.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Nitrosodi-N-methylamine-d6
CAS 17829-05-9
Molecular Formula C2 D6 N2 O
Quantity 0.05 g
Synonym Di(methyl-d3)amine, N-nitroso- (8 CI), Dimethylnitrosamine-d6, N-Nitrosodimethylamine-d6
InChI Formula InChI=1 S/C2H6N2O/c1-4(2)3-5/h1-2H3/i1D3,2D3
SMILES N(N=O)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC Name N,N-bis(trideuteriomethyl)nitrous amide
Molecular Weight (g/mol) 80.12
Formula Weight 80.0857 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity