Learn More
N-Nitrosobis(2-hydroxyethyl)-d8-amine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | Nitrosobis(2-hydroxyethyl)amine-d8 |
| CAS | 1173019-53-8 |
| Molecular Formula | C4D8H2N2O3 |
| Quantity | 0.05 g |
| Synonym | 2,2’-(Nitrosoimino)bisethanol-d8, Diethanolnitrosamine-d8, N,N-Diethanolnitrosamine-d8, N-Nitrosodiethanolamine-d8, NDELA-d8; |
| InChI Formula | InChI=1 S/C4H10N2O3/c7-3-1-6(5-9)2-4-8/h7-8 H,1-4H2/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])(O)C([2 H])([2 H])N(N=O)C([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | N,N-bis(1,1,2,2-tetradeuterio-2-hydroxy-ethyl)nitrous amide |
| Molecular Weight (g/mol) | 142.18 |
| Formula Weight | 142.1194 g/mol |
| Show More |