missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methylnicotinamide-2,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | N-Methylnicotinamide-d4 |
| CAS | 2708278-64-0 |
| Molecular Formula | C7 D4 H4 N2 O |
| Quantity | 0.05 g |
| Synonym | N-Methylnicotinamide-2,4,5,6-d4, N-Methyl-3-pyridinecarboxamide-2,4,5,6-d4, 3-(Methylcarbamoyl)pyridine-2,4,5,6-d4, N-Methylnicotinamide-d4, Nicotinic acid-d4 methylamide, N'-Methylnicotinamide-d4, N'-Methylnicotinamide-d4 |
| InChI Formula | InChI=1 S/C7H8N2O/c1-8-7(10)6-3-2-4-9-5-6/h2-5 H,1H3,(H,8,10)/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1nc([2 H])c(C(=O)NC)c([2 H])c1[2 H] |
| IUPAC Name | 2,4,5,6-tetradeuterio-N-methylpyridine-3-carboxamide |
| Molecular Weight (g/mol) | 140.176 |
| Formula Weight | 140.089 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction