missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-iso-Propyl-d7-acrylamide, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | N-iso-Propyl-d7-acrylamide |
| CAS | 1219803-32-3 |
| Molecular Formula | C6 D7 H4 N O |
| Quantity | 0.05 g |
| Synonym | 2-Propenamide, N-[1-(methyl-d3)ethyl-1,2,2,2-d4]- (ACI), N-[1-(Methyl-d3)ethyl-1,2,2,2-d4]-2-propenamide (ACI), N-iso-Propyl-d7-acrylamide, N-(Isopropyl-d7)acrylamide, N-Isopropylacrylamide-d7, N-Isopropylacrylamide-D7 (isoproyl-D7) |
| InChI Formula | InChI=1 S/C6H11NO/c1-4-6(8)7-5(2)3/h4-5 H,1H2,2-3H3,(H,7,8)/i2D3,3D3,5 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(NC(=O)C=C)C([2 H])([2 H])[2 H] |
| IUPAC Name | N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)prop-2-enamide |
| Molecular Weight (g/mol) | 120.2008 |
| Formula Weight | 120.128 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction