Learn More
n-Butyraldehyde-d8, CDN
Product Code. 30320958
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320958
Brand: CDN CDND53150.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Butanal-d8
CAS 84965-36-6
Molecular Formula C4 2H8 O
Quantity 0.05 g
Synonym Butanal-d8
InChI Formula InChI=1 S/C4H8O/c1-2-3-4-5/h4H,2-3H2,1H3/i1D3,2D2,3D2,4 D
SMILES [2 H]C(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC Name 1,2,2,3,3,4,4,4-octadeuteriobutan-1-one
Molecular Weight (g/mol) 80.16
Formula Weight 80.1077 g/mol
Percent Purity 99 atom % D, min 96% Chemical Purity