missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-S-(n-propyl-d7)-L-cysteine, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-S-(propyl-d7)-L-cysteine |
| CAS | 1331909-69-3 |
| Molecular Formula | C8 D7 H8 N O3 S |
| Quantity | 0.05 g |
| Synonym | N-Acetyl-S-(n-propyl-d7)-L-cysteine, N-Acetyl-S-(propyl-d7)-L-cysteine, N-Acetyl-S-propyl-L-cysteine-d7, N-Acetyl-S-propyl-L-cysteine-D7 (S-propyl-D7), N-Acetyl-3-(propyl-d7-thio)-L-alanine, (2 R)-2-Acetamido-3-(propylsulfanyl-d7)propanoic acid, (2 R)-2-Acetamido-3-(propylsulfanyl-d7)propanoic acid, (2 R)-2-Acetamido-3-(propylsulfanyl-d7)propanoic acid |
| InChI Formula | InChI=1 S/C8H15NO3S/c1-3-4-13-5-7(8(11)12)9-6(2)10/h7H,3-5H2,1-2H3,(H,9,10)(H,11,12)/t7-/m0/s1/i1D3,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])SC[C@H](NC(=O)C)C(=O)O |
| IUPAC Name | (2 R)-2-acetamido-3-(1,1,2,2,3,3,3-heptadeuteriopropylsulfanyl)propanoic acid |
| Molecular Weight (g/mol) | 212.318 |
| Formula Weight | 212.121 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction