Learn More
N-Acetyl-L-alanine-3,3,3-d3, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-L-alanine-3,3,3-D3 |
| CAS | 1485548-36-4 |
| Molecular Formula | C5 2H3 H6 N O3 |
| Quantity | 0.05 g |
| Synonym | N-Acetyl-L-alanine-3,3,3-D3, N-Acetyl-L-alanine-d3, (2 S)-2-Acetamido-3,3,3-trideuteriopropanoic acid, (S)-N-Acetylalanine-d3, Acetyl-L-alanine-3,3,3-d3, (S)-2-Acetamidopropionic-3,3,3-d3 acid |
| InChI Formula | InChI=1 S/C5H9NO3/c1-3(5(8)9)6-4(2)7/h3H,1-2H3,(H,6,7)(H,8,9)/t3-/m0/s1/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])[C@H](NC(=O)C)C(=O)O |
| IUPAC Name | (2 S)-2-acetamido-3,3,3-trideuteriopropanoic acid |
| Molecular Weight (g/mol) | 134.15 |
| Formula Weight | 134.0771 g/mol |
| Show More |