missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-d3-L-threonine-2,3-d2, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-L-threonine-D5 (N-acetyl,2,3-D5) |
| Molecular Formula | C6 D5 H6 N O4 |
| Quantity | 0.05 g |
| Synonym | N-Acetyl-L-threonine-D5 (N-acetyl,2,3-D5), N-(Acetyl-d3)-L-threonine-2,3-d2, N-Acetyl-L-threonine-d5, (2 S,3 R)-2-(Acet-d3-amido)-3-hydroxybutanoic-2,3-d2 acid |
| InChI Formula | InChI=1 S/C6H11NO4/c1-3(8)5(6(10)11)7-4(2)9/h3,5,8 H,1-2H3,(H,7,9)(H,10,11)/t3-,5+/m1/s1/i2D3,3 D,5 D |
| SMILES | [2 H]C([2 H])([2 H])C(=O)N[C@]([2 H])(C(=O)O)[C@@]([2 H])(C)O |
| IUPAC Name | (2 S,3 R)-2,3-dideuterio-3-hydroxy-2-[(2,2,2-trideuterioacetyl)amino]butanoic acid |
| Molecular Weight (g/mol) | 166.187 |
| Formula Weight | 166.1 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction