Learn More
mono-n-Pentyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Monopentyl Phthalate-d4 |
| CAS | 1794756-28-7 |
| Molecular Formula | C13 D4 H12 O4 |
| Quantity | 0.05 g |
| Synonym | 2,3,4,5-Tetradeuterio-6-pentoxycarbonylbenzoic acid, mono-n-Pentyl Phthalate-3,4,5,6-d4, Monopentyl Phthalate-d4, 1,2-Benzenedicarboxylic-3,4,5,6-d4 acid monopentyl ester, Phthalic acid-d4 monopentyl ester, Pentyl hydrogen phthalate-d4, Monopentyl Phthalate-3,4,5,6-D4 |
| InChI Formula | InChI=1 S/C13H16O4/c1-2-3-6-9-17-13(16)11-8-5-4-7-10(11)12(14)15/h4-5,7-8 H,2-3,6,9H2,1H3,(H,14,15)/i4D,5 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCC)c(C(=O)O)c1[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-pentoxycarbonylbenzoic acid |
| Molecular Weight (g/mol) | 240.288 |
| Formula Weight | 240.13 g/mol |
| Show More |