missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mono-n-Dodecyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | Monododecyl Phthalate-3,4,5,6-D4 |
| CAS | 2732763-22-1 |
| Molecular Formula | C20 D4 H26 O4 |
| Quantity | 0.05 g |
| Synonym | 1-Dodecyl 1,2-benzenedicarboxylate-3,4,5,6-d4, Phthalic-d4 acid monododecyl ester, 2-(Dodecyloxycarbonyl)benzoic-3,4,5,6-d4 acid, Dodecyl hydrogen phthalate, Dodecyl phthalate-d4, Lauryl phthalate-d4, Monododecyl Phthalate-3,4,5,6-D4, Monolauryl phthalate-3,4,5,6-d4 |
| InChI Formula | InChI=1 S/C20H30O4/c1-2-3-4-5-6-7-8-9-10-13-16-24-20(23)18-15-12-11-14-17(18)19(21)22/h11-12,14-15 H,2-10,13,16H2,1H3,(H,21,22)/i11D,12 D,14 D,15 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCCCCCCCCC)c(C(=O)O)c1[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-dodecoxycarbonylbenzoic acid |
| Molecular Weight (g/mol) | 338.4745 |
| Formula Weight | 338.2395 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction