missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Proline-2,5,5-d3-N-FMOC, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-proline-2,5,5-D3 |
| Molecular Formula | C20 D3 H16 N O4 |
| Quantity | 0.05 g |
| Synonym | L-Proline-2,5,5-d3-N-FMOC, 1-(9 H-Fluoren-9-ylmethyl) (2 S)-1,2-pyrrolidinedicarboxylate-2,5,5-d3, (2 S)-Pyrrolidine-1,2-dicarboxylic-2,5,5-d3 acid 1-(9 H-fluoren-9-ylmethyl) ester, (S)-N-(9-Fluorenylmethoxycarbonyl)-proline-2,5,5-d3, (S)-N-FMOC-proline-2,5,5-d3, N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-proline-2,5,5-D3, N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-proline-2,5,5-D3, N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-proline-2,5,5-D3 |
| InChI Formula | InChI=1 S/C20H19NO4/c22-19(23)18-10-5-11-21(18)20(24)25-12-17-15-8-3-1-6-13(15)14-7-2-4-9-16(14)17/h1-4,6-9,17-18 H,5,10-12H2,(H,22,23)/t18-/m0/s1/i11D2,18 D |
| SMILES | [2 H]C1([2 H])CC[C@]([2 H])(N1C(=O)OCC2c3ccccc3c4ccccc24)C(=O)O |
| IUPAC Name | (2 S)-2,5,5-trideuterio-1-(9 H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carboxylic acid |
| Molecular Weight (g/mol) | 340.3877 |
| Formula Weight | 340.1502 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction