missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-3-(4-Hydroxy-3-methoxy-d3-phenyl)alanine H2O, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 3-O-Methyl-L-DOPA-d3 Monohydrate |
| Molecular Formula | C10 H10 D3 N O4 . H2 O |
| Quantity | 0.05 g |
| Synonym | 3-(Methoxy-d3)-L-tyrosine Monohydrate, 3-(Methoxy-d3)-4-hydroxyphenylalanine Monohydrate, 3-(Methoxy-d3)-DOPA Monohydrate, 3-(Methoxy-d3)-L-tyrosine Monohydrate, 3-O-(Methyl-d3)dopa Monohydrate, L-3-(4-Hydroxy-3-methoxy-d3-phenyl)alanine H2O, L-3-(4-Hydroxy-3-methoxy-d3-phenyl)alanine H2O, L-3-(4-Hydroxy-3-methoxy-d3-phenyl)alanine H2O, L-3-(4-Hydroxy-3-methoxy-d3-phenyl)alanine H2O |
| InChI Formula | InChI=1 S/C10H13NO4.H2O/c1-15-9-5-6(2-3-8(9)12)4-7(11)10(13)14;/h2-3,5,7,12 H,4,11H2,1H3,(H,13,14);1H2/t7-;/m0./s1/i1D3; |
| SMILES | O.[2 H]C([2 H])([2 H])Oc1cc(C[C@H](N)C(=O)O)ccc1O |
| IUPAC Name | (2 S)-2-amino-3-[4-hydroxy-3-(trideuteriomethoxy)phenyl]propanoic acid;hydrate |
| Molecular Weight (g/mol) | 232.2483 |
| Formula Weight | 232.1139 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction