missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Forchlorfenuron-d5 (phenyl-d5), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Forchlorfenuron D5 (phenyl D5) |
| CAS | 1398065-87-6 |
| Molecular Formula | C12 2H5 H5 Cl N3 O |
| Quantity | 0.05 g |
| Synonym | Forchlorfenuron D5 (phenyl D5), N-(2-Chloro-4-pyridinyl)-N'-(phenyl-d5)urea, 1-(2-Chloro-4-pyridyl)-3-(phenyl-d5)urea |
| InChI Formula | InChI=1 S/C12H10ClN3O/c13-11-8-10(6-7-14-11)16-12(17)15-9-4-2-1-3-5-9/h1-8 H,(H2,14,15,16,17)/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(NC(=O)Nc2ccnc(Cl)c2)c([2 H])c1[2 H] |
| IUPAC Name | 1-(2-chloropyridin-4-yl)-3-(2,3,4,5,6-pentadeuteriophenyl)urea |
| Molecular Weight (g/mol) | 252.71 |
| Formula Weight | 252.0826 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction