missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diquat-d4 Dibromide (ethylene-d4), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Diquat-d4 Dibromide |
| Molecular Formula | C12 D4 H8 N2 . 2 Br |
| Quantity | 0.05 g |
| Synonym | Diquat dibromide D4, Diquat Dibromide D4, Diquat-d4 Dibromide, Diquat-d4 (ethylene-d4) dibromide |
| InChI Formula | InChI=1 S/C12H12N2.2 BrH/c1-3-7-13-9-10-14-8-4-2-6-12(14)11(13)5-1;;/h1-8 H,9-10H2;2*1 H/q+2;;/p-2/i9D2,10D2;; |
| SMILES | [Br-].[Br-].[2 H]C1([2 H])[n+]2 ccccc2c3cccc[n+]3C1([2 H])[2 H] |
| IUPAC Name | 6,6,7,7-tetradeuteriodipyrido[2,1-a:2',1'-d]pyrazine-5,8-diium;dibromide |
| Molecular Weight (g/mol) | 348.0697 |
| Formula Weight | 345.9618 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction