missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-propyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-propyl ester D4 |
| CAS | 358731-29-0 |
| Molecular Formula | C14 2H4 H14 O4 |
| Quantity | 0.05 g |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dipropyl ester (9 CI), Di-n-propyl Phthalate-d4, Phthalic acid, bis-propyl ester D4 |
| InChI Formula | InChI=1 S/C14H18O4/c1-3-9-17-13(15)11-7-5-6-8-12(11)14(16)18-10-4-2/h5-8 H,3-4,9-10H2,1-2H3/i5D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCCC)c(C(=O)OCCC)c1[2 H] |
| IUPAC Name | dipropyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 254.32 |
| Formula Weight | 254.1456 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction