missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desethylterbuthylazine-d9 (tert-butyl-d9), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Desethyl Terbuthylazine-d9 |
| CAS | 1219798-52-3 |
| Molecular Formula | C7 2H9 H3 Cl N5 |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C7H12ClN5/c1-7(2,3)13-6-11-4(8)10-5(9)12-6/h1-3H3,(H3,9,10,11,12,13)/i1D3,2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])C(Nc1nc(N)nc(Cl)n1)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 6-chloro-4-N-[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]-1,3,5-triazine-2,4-diamine |
| Molecular Weight (g/mol) | 210.71 |
| Formula Weight | 210.1346 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction